Gene Transcripts

Gene Symbol
Klri1
Entrez Gene ID
503550
Gene Name
killer cell lectin-like receptor family I member 1
Gene Aliases
Chromosome Location
Chr. 6: 129696118 - 129720610 on Build GRCm38
UniGene ID
Mm.376476
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012520.2 NP_001012538.2 1 131
GenBank mRNA AY324874.1 - 1 131

View Full Product Details