Gene Transcripts

Gene Symbol
Defb25
Entrez Gene ID
654459
Gene Name
defensin beta 25
Gene Aliases
BD-25, OTTMUSG00000015862, mBD-25
Chromosome Location
Chr. 2: 152622356 - 152623053 on Build GRCm38
UniGene ID
Mm.358531
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039122.1 NP_001034211.1 1, 2 91
GenBank mRNA BC132662.1 - 1, 2 87
DQ012036.1 - 1, 2 91

View Full Product Details