Gene Transcripts

Gene Symbol
Myo3a
Entrez Gene ID
667663
Gene Name
myosin IIIA
Gene Aliases
9030416P08Rik
Chromosome Location
Chr. 2: 22227503 - 22423420 on Build GRCm38
UniGene ID
Mm.330033
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148413.3 NP_680779.3 1405
GenBank mRNA AY101368.1 - 1248

View Full Product Details