Gene Transcripts

Gene Symbol
Prl4a1
Entrez Gene ID
24656
Gene Name
prolactin family 4, subfamily a, member 1
Gene Aliases
Plpa, Prlpa
Chromosome Location
Chr. 17: 38710401 - 38718343 on Build Rnor_6.0
UniGene ID
Rn.11230
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017036.2 NP_058732.2 5 642
GenBank mRNA BC097332.1 - 5 642
FQ228460.1 - 5 677
M13750.1 - 5 625

View Full Product Details