Gene Transcripts

Gene Symbol
Nfia
Entrez Gene ID
25492
Gene Name
nuclear factor I/A
Gene Aliases
Chromosome Location
Chr. 5: 116421895 - 116750381 on Build Rnor_6.0
UniGene ID
Rn.10550
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012988.2 NP_037120.1 4 911
GenBank mRNA AB060652.1 - 819
AF112455.1 - 991
AY572794.1 - 4 819
D78017.1 - 4 911
D78018.1 - 3 840
D78020.1 - 4 333
X13167.1 - 4 753

View Full Product Details