Gene Transcripts

Gene Symbol
Ctsj
Entrez Gene ID
29174
Gene Name
cathepsin J
Gene Aliases
Ctsp
Chromosome Location
Chr. 17: 4105890 - 4110538 on Build Rnor_6.0
UniGene ID
Rn.34875
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017121.1 NP_058817.1 2 177
XM_008771439.1 XP_008769661.1 1 95
GenBank mRNA AF310623.2 - 2 177
BC097263.1 - 2 160 7460052

View Full Product Details