Gene Transcripts
- Gene Symbol
- Klrc1
- Entrez Gene ID
- 29683
- Gene Name
- killer cell lectin-like receptor subfamily C, member 1
- Gene Aliases
- rNKG2A
- Chromosome Location
- Chr. 4: 163453435 - 163463718 on Build Rnor_6.0
- UniGene ID
- Rn.9895
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF021350.1 |
- |
2, 3 |
279 |
|
View Full Product Details