Gene Transcripts
- Gene Symbol
- SNRPA1
- Entrez Gene ID
- 6627
- Gene Name
- small nuclear ribonucleoprotein polypeptide A'
- Gene Aliases
- Lea1
- Chromosome Location
- Chr. 15: 101281510 - 101295282 on Build GRCh38
- UniGene ID
- Hs.528763
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003090.3 |
NP_003081.2
|
2, 3 |
331 |
|
NR_135508.1 |
|
2, 3 |
331 |
|
GenBank mRNA |
AK223070.1 |
- |
2, 3 |
288 |
|
AK223110.1 |
- |
2, 3 |
253 |
|
AK290078.1 |
- |
2, 3 |
287 |
|
AK312200.1 |
- |
2, 3 |
262 |
|
AL538391.3 |
- |
2, 3 |
330 |
|
BC022816.1 |
- |
2, 3 |
284 |
4904205 |
BC067846.1 |
- |
2, 3 |
300 |
30330402 |
BC071717.1 |
- |
1 |
239 |
4514975 |
X13482.1 |
- |
2, 3 |
283 |
|
View Full Product Details