Gene Transcripts

Gene Symbol
Adam3a
Entrez Gene ID
57021
Gene Name
ADAM metallopeptidase domain 3A
Gene Aliases
Adam3, tMDCI
Chromosome Location
Chr. 16: 72086878 - 72136685 on Build Rnor_6.0
UniGene ID
Rn.42918
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020302.1 NP_064698.1 13, 14 1455
GenBank mRNA BC078839.1 - 13, 14 1361
Y07903.1 - 13, 14 1455

View Full Product Details