Gene Transcripts

Gene Symbol
SPAST
Entrez Gene ID
6683
Gene Name
spastin
Gene Aliases
ADPSP, FSP2, SPG4
Chromosome Location
Chr. 2: 32063551 - 32157637 on Build GRCh38
UniGene ID
Hs.468091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014946.3 NP_055761.2 6 1195
NM_199436.1 NP_955468.1 5 1099
XM_005264516.4 XP_005264573.1 6 971
XM_011533067.2 XP_011531369.1 6 1255
XM_017004777.1 XP_016860266.1 5 1156
XM_017004778.1 XP_016860267.1 5 1159
GenBank mRNA AB029006.1 - 5 1099
AJ246001.1 - 6 1099
BC150260.1 - 5 1099 9007202

View Full Product Details