Gene Transcripts

Gene Symbol
Lphn1
Entrez Gene ID
65096
Gene Name
latrophilin 1
Gene Aliases
CL1BA
Chromosome Location
Chr. 19: 25029037 - 25069146 on Build Rnor_6.0
UniGene ID
Rn.10776
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022962.1 NP_075251.1 6 979
GenBank mRNA AF081144.1 - 5 964
AF081145.1 - 5 964
AF081146.1 - 6 979
AF081147.1 - 6 979
AF111099.1 - 7 1305
U72487.1 - 5 781
U78105.1 - 5 1106

View Full Product Details