Gene Transcripts
- Gene Symbol
- Rpl37
- Entrez Gene ID
- 81770
- Gene Name
- ribosomal protein L37
- Gene Aliases
- Chromosome Location
- Chr. 2: 54832634 - 54834587 on Build Rnor_6.0
- UniGene ID
- Rn.129486
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031106.1 |
NP_112368.1
|
3, 4 |
256 |
|
GenBank mRNA |
BC059132.1 |
- |
3, 4 |
253 |
|
BC069173.1 |
- |
3, 4 |
238 |
7103990 |
FQ210660.1 |
- |
3, 4 |
259 |
|
FQ210874.1 |
- |
3, 4 |
259 |
|
FQ217102.1 |
- |
3, 4 |
259 |
|
FQ217508.1 |
- |
3, 4 |
249 |
|
FQ220738.1 |
- |
3, 4 |
259 |
|
FQ221496.1 |
- |
3, 4 |
249 |
|
FQ221700.1 |
- |
3, 4 |
249 |
|
FQ222983.1 |
- |
3, 4 |
259 |
|
FQ223019.1 |
- |
3, 4 |
259 |
|
FQ223296.1 |
- |
3, 4 |
264 |
|
FQ223471.1 |
- |
3, 4 |
249 |
|
S79981.1 |
- |
3, 4 |
256 |
|
X66369.1 |
- |
3, 4 |
256 |
|
- Gene Symbol
- LOC100360841
- Entrez Gene ID
- 100360841
- Gene Name
- ribosomal protein L37-like
- Gene Aliases
- Chromosome Location
- Chr. 9: 69790835 - 69791192 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- RGD1561310
- Entrez Gene ID
- 498744
- Gene Name
- similar to ribosomal protein L37
- Gene Aliases
- Chromosome Location
- Chr. 17: 36438176 - 36438782 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details