Gene Transcripts

Gene Symbol
Srp54a
Entrez Gene ID
116650
Gene Name
signal recognition particle 54A
Gene Aliases
Srp54
Chromosome Location
Chr. 6: 75996631 - 76035763 on Build Rnor_6.0
UniGene ID
Rn.24880
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053871.1 NP_446323.1 13 1388
XM_006240097.2 XP_006240159.1 13 1400
GenBank mRNA BC079117.1 - 13 1388
FQ211580.1 - 13 1399
FQ225417.1 - 13 1403

View Full Product Details