Gene Transcripts

Gene Symbol
Fat3
Entrez Gene ID
191571
Gene Name
FAT atypical cadherin 3
Gene Aliases
Chromosome Location
Chr. 8: 14417039 - 15011596 on Build Rnor_6.0
UniGene ID
Rn.210528
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138544.1 NP_612553.1 2 1826
GenBank mRNA AB076401.1 - 2 1826

View Full Product Details