Gene Transcripts

Gene Symbol
Zfp91
Entrez Gene ID
246282
Gene Name
zinc finger protein 91
Gene Aliases
Chromosome Location
Chr. 1: 229602499 - 229640917 on Build Rnor_6.0
UniGene ID
Rn.1772
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169120.1 NP_001162591.1 10 1203
XM_006231127.2 XP_006231189.1 10 1457

View Full Product Details