Gene Transcripts

Gene Symbol
Prl8a5
Entrez Gene ID
286889
Gene Name
prolactin family 8, subfamily a, member 5
Gene Aliases
Chromosome Location
Chr. 17: 39411716 - 39417929 on Build Rnor_6.0
UniGene ID
Rn.3757
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173110.1 NP_775133.1 6 616
XM_006253907.1 XP_006253969.1 6 626
XM_006253908.1 XP_006253970.1 6 582
GenBank mRNA BC097274.1 - 5 563
BC158577.1 - 6 610 7461869
M76537.1 - 6 593
U09099.1 - 6 616

View Full Product Details