Gene Transcripts
- Gene Symbol
- Rhot2
- Entrez Gene ID
- 287156
- Gene Name
- ras homolog family member T2
- Gene Aliases
- Miro2
- Chromosome Location
- Chr. 10: 15205945 - 15211739 on Build Rnor_6.0
- UniGene ID
- Rn.8882
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181823.1 |
NP_861544.1
|
11 |
1064 |
|
GenBank mRNA |
AY303973.1 |
- |
11 |
1064 |
|
View Full Product Details