Gene Transcripts

Gene Symbol
Zbtb4
Entrez Gene ID
287441
Gene Name
zinc finger and BTB domain containing 4
Gene Aliases
Chromosome Location
Chr. 10: 56367669 - 56388294 on Build Rnor_6.0
UniGene ID
Rn.15836
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246843.2 XP_006246905.1 4 2930
XM_006246844.2 XP_006246906.1 3 2754

View Full Product Details