Gene Transcripts

Gene Symbol
Mrpl27
Entrez Gene ID
287635
Gene Name
mitochondrial ribosomal protein L27
Gene Aliases
Chromosome Location
Chr. 10: 82375572 - 82381332 on Build Rnor_6.0
UniGene ID
Rn.18445
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105831.1 NP_001099301.1 4 298
GenBank mRNA FQ212021.1 - 4 307

View Full Product Details