Gene Transcripts

Gene Symbol
Hhip
Entrez Gene ID
291936
Gene Name
Hedgehog-interacting protein
Gene Aliases
RGD1564108
Chromosome Location
Chr. 19: 31525134 - 31614487 on Build Rnor_6.0
UniGene ID
Rn.204425
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191817.1 NP_001178746.1 9 1528

View Full Product Details