Gene Transcripts

Gene Symbol
Olr23
Entrez Gene ID
293085
Gene Name
olfactory receptor 23
Gene Aliases
ratchr1-142755772-142756767_ORF
Chromosome Location
Chr. 1: 150067242 - 150068237 on Build Rnor_6.0
UniGene ID
Rn.141947
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000119.1 NP_001000119.1 1 794

View Full Product Details