Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134428.1 | NP_001127900.1 | 2 | 193 | |
NM_001134429.1 | NP_001127901.1 | 2 | 229 | ||
XM_006230756.1 | XP_006230818.1 | 2 | 281 | ||
XM_006230757.1 | XP_006230819.1 | 2 | 245 | ||
XM_006230758.2 | XP_006230820.1 | 2 | 683 | ||
GenBank mRNA | BG669308.1 | - | 2 | 193 | |
H35224.1 | - | 2 | 182 |