Gene Transcripts

Gene Symbol
Taar4
Entrez Gene ID
294122
Gene Name
trace amine-associated receptor 4
Gene Aliases
Ta2, taR-2, taR-4
Chromosome Location
Chr. 1: 22558787 - 22559830 on Build Rnor_6.0
UniGene ID
Rn.138138
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175583.1 NP_783173.1 1 997

View Full Product Details