Gene Transcripts

Gene Symbol
Znfx1
Entrez Gene ID
296384
Gene Name
zinc finger, NFX1-type containing 1
Gene Aliases
Ab1-133, RGD1306344
Chromosome Location
Chr. 3: 163787101 - 163847671 on Build Rnor_6.0
UniGene ID
Rn.202897
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047860.1 NP_001041325.1 7 2235
XM_006235641.1 XP_006235703.1 4 2333
XM_006235642.2 XP_006235704.1 4 2073
XM_006235643.2 XP_006235705.1 4 1981
GenBank mRNA AY325141.1 - 7 2235

View Full Product Details