Gene Transcripts

Gene Symbol
Zfp93
Entrez Gene ID
296399
Gene Name
zinc finger protein 93
Gene Aliases
Chromosome Location
Chr. 3: 165679307 - 165700489 on Build Rnor_6.0
UniGene ID
Rn.133759
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106546.1 NP_001100016.1 5 1130

View Full Product Details