Gene Transcripts

Gene Symbol
ZNF17
Entrez Gene ID
7565
Gene Name
zinc finger protein 17
Gene Aliases
HPF3, KOX10
Chromosome Location
Chr. 19: 57409252 - 57421939 on Build GRCh38
UniGene ID
Hs.729403
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006959.2 NP_008890.2 3 2086
XM_017027205.1 XP_016882694.1 4 4036
GenBank mRNA AB075827.1 - 1 1693
AK097123.1 - 1 1372
AK098183.1 - 3 2068
AK127948.1 - 4 2125
AK128471.1 - 1 2291

View Full Product Details