Gene Transcripts

Gene Symbol
Sec61b
Entrez Gene ID
298068
Gene Name
Sec61 beta subunit
Gene Aliases
Chromosome Location
Chr. 5: 63192322 - 63200532 on Build Rnor_6.0
UniGene ID
Rn.103030
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106654.2 NP_001100124.1 4 466
GenBank mRNA BC167110.1 - 4 466
FQ210322.1 - 4 454
FQ217838.1 - 4 419
FQ221908.1 - 4 410

View Full Product Details