Gene Transcripts

Gene Symbol
Sav1
Entrez Gene ID
299116
Gene Name
salvador family WW domain containing protein 1
Gene Aliases
Chromosome Location
Chr. 6: 92376470 - 92398678 on Build Rnor_6.0
UniGene ID
Rn.14087
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097581.1 NP_001091050.1 2 636
GenBank mRNA AM261215.1 - 2 636
BC161803.1 - 2 483 8371277

View Full Product Details