Gene Transcripts

Gene Symbol
Sf3a2
Entrez Gene ID
299620
Gene Name
splicing factor 3a, subunit 2
Gene Aliases
Chromosome Location
Chr. 7: 11777826 - 11784287 on Build Rnor_6.0
UniGene ID
Rn.102569
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011986.1 NP_001011986.1 2, 3 199
XM_006240885.2 XP_006240947.1 1, 2 156
XM_006240886.2 XP_006240948.1 2, 3 453
GenBank mRNA BC079320.1 - 2, 3 199

View Full Product Details