Gene Transcripts

Gene Symbol
Gxylt1
Entrez Gene ID
300173
Gene Name
glucoside xylosyltransferase 1
Gene Aliases
Glt8d3
Chromosome Location
Chr. 7: 134447578 - 134495836 on Build Rnor_6.0
UniGene ID
Rn.14826
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100887.1 NP_001094357.1 3 508
XM_006242192.2 XP_006242254.1 2 416
GenBank mRNA AY623036.1 - 1 111
BC167068.1 - 2 262 8363586

View Full Product Details