Gene Transcripts

Gene Symbol
Myg1
Entrez Gene ID
300258
Gene Name
melanocyte proliferating gene 1
Gene Aliases
C12orf10
Chromosome Location
Chr. 7: 143929662 - 143936865 on Build Rnor_6.0
UniGene ID
Rn.1226
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005545.1 NP_001005545.1 6 934
GenBank mRNA BC082112.1 - 6 934
FQ213994.1 - 4 999
FQ230067.1 - 6 964

View Full Product Details