Gene Transcripts

Gene Symbol
Iqch
Entrez Gene ID
300776
Gene Name
IQ motif containing H
Gene Aliases
RGD1310670
Chromosome Location
Chr. 8: 68332526 - 68525980 on Build Rnor_6.0
UniGene ID
Rn.218384
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106830.1 NP_001100300.1 17 2455
XM_008766234.1 XP_008764456.1 11 1971
XM_008766235.1 XP_008764457.1 11 1863
XM_008766236.1 XP_008764458.1 12 2055
XM_008766237.1 XP_008764459.1 17 2556

View Full Product Details