Gene Transcripts

Gene Symbol
Mrpl14
Entrez Gene ID
301250
Gene Name
mitochondrial ribosomal protein L14
Gene Aliases
L14mt, L32mt, MRP-L14, MRP-L32
Chromosome Location
Chr. 9: 17687622 - 17698588 on Build Rnor_6.0
UniGene ID
Rn.35669
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106890.2 NP_001100360.1 3 422
NM_001271131.1 NP_001258060.1 3 378
XM_006244512.2 XP_006244574.1 3 612
XM_008766849.1 XP_008765071.1 4 545
XM_008766850.1 XP_008765072.1 3 435
GenBank mRNA FM100935.1 - 3 352

View Full Product Details