Gene Transcripts
- Gene Symbol
- Lrrc16a
- Entrez Gene ID
- 306941
- Gene Name
- leucine rich repeat containing 16A
- Gene Aliases
- Lrrc16
- Chromosome Location
- Chr. 17: 43050961 - 43339403 on Build Rnor_6.0
- UniGene ID
- Rn.40387
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001191692.1 |
NP_001178621.1
|
19, 20 |
1497 |
|
XM_008771640.1 |
XP_008769862.1
|
11, 12 |
1096 |
|
XM_008771641.1 |
XP_008769863.1
|
11, 12 |
1096 |
|
XM_008771642.1 |
XP_008769864.1
|
10, 11 |
1084 |
|
XM_008771643.1 |
XP_008769865.1
|
11, 12 |
1096 |
|
XM_008771644.1 |
XP_008769866.1
|
10, 11 |
1084 |
|
XM_008771645.1 |
XP_008769867.1
|
11, 12 |
1096 |
|
XM_008771646.1 |
XP_008769868.1
|
11, 12 |
1096 |
|
XM_008771647.1 |
XP_008769869.1
|
10, 11 |
1083 |
|
XM_008771648.1 |
XP_008769870.1
|
11, 12 |
1096 |
|
XM_008771649.1 |
XP_008769871.1
|
11, 12 |
1096 |
|
XM_008771650.1 |
XP_008769872.1
|
11, 12 |
1096 |
|
XM_008771651.1 |
XP_008769873.1
|
11, 12 |
1096 |
|
XM_008771652.1 |
XP_008769874.1
|
11, 12 |
1096 |
|
View Full Product Details