Gene Transcripts

Gene Symbol
Pcdhb22
Entrez Gene ID
307486
Gene Name
protocadherin beta 22
Gene Aliases
pcdh-T5
Chromosome Location
Chr. 18: 30592787 - 30595456 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001056235.5 XP_001056235.1 1 418

View Full Product Details