Gene Transcripts

Gene Symbol
Irx6
Entrez Gene ID
307715
Gene Name
iroquois homeobox 6
Gene Aliases
RGD1564830
Chromosome Location
Chr. 19: 15726654 - 15733412 on Build Rnor_6.0
UniGene ID
Rn.151302
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102413.1 NP_001095883.1 2, 3 1032
XM_006255173.2 XP_006255235.1 2, 3 1011

View Full Product Details