Gene Transcripts

Gene Symbol
Slitrk3
Entrez Gene ID
310519
Gene Name
SLIT and NTRK-like family, member 3
Gene Aliases
Chromosome Location
Chr. 2: 170449182 - 170461513 on Build Rnor_6.0
UniGene ID
Rn.68333
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107683.1 NP_001101153.1 2 487
XM_006232490.2 XP_006232552.1 2 757
XM_008761091.1 XP_008759313.1 2 229

View Full Product Details