Gene Transcripts

Gene Symbol
Sprr3
Entrez Gene ID
310575
Gene Name
small proline-rich protein 3
Gene Aliases
Chromosome Location
Chr. 2: 192641472 - 192643942 on Build Rnor_6.0
UniGene ID
Rn.230057
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107686.1 NP_001101156.1 1 535
XM_008761142.1 XP_008759364.1 2 585

View Full Product Details