Gene Transcripts

Gene Symbol
Frrs1
Entrez Gene ID
310810
Gene Name
ferric-chelate reductase 1
Gene Aliases
Sdfr2
Chromosome Location
Chr. 2: 220415567 - 220469420 on Build Rnor_6.0
UniGene ID
Rn.162616
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761474.1 XP_008759696.1 13 1606
GenBank mRNA FQ210266.1 - 13 1543

View Full Product Details