Gene Transcripts

Gene Symbol
Ttc17
Entrez Gene ID
311224
Gene Name
tetratricopeptide repeat domain 17
Gene Aliases
Chromosome Location
Chr. 3: 83198832 - 83306619 on Build Rnor_6.0
UniGene ID
Rn.207415
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107752.2 NP_001101222.2 5 594
XM_006234582.2 XP_006234644.1 5 623
GenBank mRNA BC168714.1 - 5 594

View Full Product Details