Gene Transcripts

Gene Symbol
Pla2g4d
Entrez Gene ID
691905
Gene Name
phospholipase A2, group IVD (cytosolic)
Gene Aliases
Chromosome Location
Chr. 3: 112040827 - 112061847 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001080051.5 XP_001080051.2 4 368

View Full Product Details