Gene Transcripts

Gene Symbol
Klhdc10
Entrez Gene ID
312199
Gene Name
kelch domain containing 10
Gene Aliases
RGD1309792
Chromosome Location
Chr. 4: 57715979 - 57773800 on Build Rnor_6.0
UniGene ID
Rn.213729
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017456.1 NP_001017456.1 6 857
XM_006236201.2 XP_006236263.1 7 1013
GenBank mRNA BC085351.1 - 6 857

View Full Product Details