Gene Transcripts

Gene Symbol
Ythdf2
Entrez Gene ID
313053
Gene Name
YTH N(6)-methyladenosine RNA binding protein 2
Gene Aliases
Chromosome Location
Chr. 5: 150364490 - 150390173 on Build Rnor_6.0
UniGene ID
Rn.21737
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047099.1 NP_001040564.1 5 711
GenBank mRNA AB259152.1 - 5 711
BC166407.1 - 4 626 7453874

View Full Product Details