Gene Transcripts

Gene Symbol
Zmym4
Entrez Gene ID
313598
Gene Name
zinc finger, MYM-type 4
Gene Aliases
Zfp262
Chromosome Location
Chr. 5: 144878212 - 144996431 on Build Rnor_6.0
UniGene ID
Rn.3185
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107982.1 NP_001101452.1 11 2077
XM_006238873.1 XP_006238935.1 11 2072

View Full Product Details