Gene Transcripts

Gene Symbol
Insm2
Entrez Gene ID
314131
Gene Name
insulinoma-associated 2
Gene Aliases
Chromosome Location
Chr. 6: 76382181 - 76386230 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006240111.2 XP_006240173.1 1 2334

View Full Product Details