Gene Transcripts

Gene Symbol
MKNK1
Entrez Gene ID
8569
Gene Name
MAP kinase interacting serine/threonine kinase 1
Gene Aliases
MNK1
Chromosome Location
Chr. 1: 46557407 - 46607140 on Build GRCh38
UniGene ID
Hs.371594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135553.2 NP_001129025.1 7 721
NM_003684.5 NP_003675.2 7 721
NM_198973.3 NP_945324.1 7 679
NR_024174.2 6 647
NR_024176.2 7 748
XM_006711000.1 XP_006711063.1 7 674
XM_006711001.2 XP_006711064.1 8 924
XM_006711002.2 XP_006711065.1 7 6149
XM_006711003.1 XP_006711066.1 8 735
XM_006711006.3 XP_006711069.1 6 3464
XM_006711007.1 XP_006711070.1 6 566
XM_006711009.3 XP_006711072.1 6 6051
XM_006711013.1 XP_006711076.1 6 599
XM_006711014.1 XP_006711077.1 6 590
XM_006711015.1 XP_006711078.1 7 661
XM_017002652.1 XP_016858141.1 7 6397
XM_017002653.1 XP_016858142.1 8 1022
XM_017002654.1 XP_016858143.1 8 3874
XM_017002655.1 XP_016858144.1 8 6495
XM_017002656.1 XP_016858145.1 8 6294
XM_017002657.1 XP_016858146.1 7 6196
XM_017002658.1 XP_016858147.1 7 6397
XM_017002659.1 XP_016858148.1 7 693
XM_017002660.1 XP_016858149.1 7 6149
XM_017002661.1 XP_016858150.1 8 914
XM_017002662.1 XP_016858151.1 8 735
XM_017002663.1 XP_016858152.1 8 6495
XM_017002664.1 XP_016858153.1 6 6075
XM_017002665.1 XP_016858154.1 6 566
XM_017002666.1 XP_016858155.1 7 3550
XM_017002667.1 XP_016858156.1 5 1126
XM_017002668.1 XP_016858157.1 7 6397
GenBank mRNA AB000409.1 - 7 664
AB451300.1 - 6 477
AB451432.1 - 6 477
AK096423.1 - 8 766
AK290456.1 - 7 641
AK298839.1 - 6 621
AK310566.1 - 6 566
AY355461.1 - 6 477
BC002755.2 - 7 641 3629765
BG722049.1 - 7 683
BX538193.1 - 5 471

View Full Product Details