Gene Transcripts

Gene Symbol
Wdr24
Entrez Gene ID
360497
Gene Name
WD repeat domain 24
Gene Aliases
RGD1304803
Chromosome Location
Chr. 10: 15189772 - 15195860 on Build Rnor_6.0
UniGene ID
Rn.20330
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191084.1 NP_001178013.1 3 1312
XM_008767587.1 XP_008765809.1 3 2618

View Full Product Details