Gene Transcripts

Gene Symbol
Sel1l3
Entrez Gene ID
360945
Gene Name
sel-1 suppressor of lin-12-like 3 (C. elegans)
Gene Aliases
RGD1310366, RGD1562860
Chromosome Location
Chr. 14: 60122984 - 60229565 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006251033.2 XP_006251095.1 8 3692
XM_341223.7 XP_341224.5 8 1488

View Full Product Details