Gene Transcripts

Gene Symbol
KSR1
Entrez Gene ID
8844
Gene Name
kinase suppressor of ras 1
Gene Aliases
KSR, RSU2
Chromosome Location
Chr. 17: 27456441 - 27626438 on Build GRCh38
UniGene ID
Hs.133534
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014238.1 NP_055053.1 11, 12 1513
XM_011525429.2 XP_011523731.1 10, 11 1665
XM_011525430.2 XP_011523732.1 10, 11 1667
XM_011525431.2 XP_011523733.1 10, 11 1667
XM_017025268.1 XP_016880757.1 10, 11 1665
XM_017025269.1 XP_016880758.1 10, 11 1665
XM_017025270.1 XP_016880759.1 9, 10 1400
XM_017025271.1 XP_016880760.1 11, 12 2372
XM_017025272.1 XP_016880761.1 11, 12 2355
XM_017025273.1 XP_016880762.1 10, 11 2178
XM_017025274.1 XP_016880763.1 10, 11 1381
XM_017025275.1 XP_016880764.1 10, 11 1368
XM_017025276.1 XP_016880765.1 11, 12 2320
XM_017025277.1 XP_016880766.1 6, 7 1063
XM_017025278.1 XP_016880767.1 6, 7 1073
GenBank mRNA BC035488.1 - 12, 13 2585
BC042106.1 - 11, 12 1512
U43586.1 - 7, 8 753

View Full Product Details