Gene Transcripts

Gene Symbol
NR1I2
Entrez Gene ID
8856
Gene Name
nuclear receptor subfamily 1 group I member 2
Gene Aliases
BXR, ONR1, PAR, PAR1, PAR2, PARq, PRR, PXR, SAR, SXR
Chromosome Location
Chr. 3: 119780484 - 119818485 on Build GRCh38
UniGene ID
Hs.7303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003889.3 NP_003880.3 2 1995
NM_022002.2 NP_071285.1 2 321
NM_033013.2 NP_148934.1 2 1995
GenBank mRNA AB307701.1 - 1 156
AF061056.1 - 2 459
AF084644.1 - 2 332
AF084645.1 - 2 435
AJ009936.1 - 3 1995
AJ009937.1 - 3 1995
AK223456.1 - 2 440
AK310789.1 - 2 444
AY091855.1 - 2 738
BC017304.2 - 2 395 5087401
HQ692837.1 - 2 273
HQ709177.1 - 2 273

View Full Product Details